4-Phenoxythiobenzamide
Catalog # FLUO021342-1G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
CAS Number:730971-68-3
Formula:C₁₃H₁₁NOS
MDL Number:MFCD04973319