Order Entry
Germany
Orders LinkContactUsLinkComponent
2-Hydroxy-4-nitrobenzonitrile

2-Hydroxy-4-nitrobenzonitrile

Marketsource
Supplier:   CHEMPUR
2-Hydroxy-4-nitrobenzonitrile
Supplier:  CHEMPUR
Marketsource
Certificates
2-Hydroxy-4-nitrobenzonitrile

2 Options Available Below

Product Details & Documents
  • CAS Number:39835-14-8
  • Formula:C₇H₄N₂O₃
  • MDL Number:MFCD02258875
  • Molecular Weight:164.1183 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • 1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 223,00 €
    223,00 €
    1 * 5 g
       223,00 €

    Specifications:
    Marketsource
    Marketsource