Order Entry
Germany
ContactUsLinkComponent
 
4-(1,3-Oxazol-2-yl)aniline hydrochloride
Catalog # COBBHC-3349-1G
Supplier:  COMBI-BLOCKS
CAS Number:  
4-(1,3-Oxazol-2-yl)aniline hydrochloride
Catalog # COBBHC-3349-1G
Supplier:  COMBI-BLOCKS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
CAS Number:1351659-13-6
Formula:C₉H₉ClN₂O
MDL Number:MFCD19237193