5-Benzylmercaptotetrazole (BMT, BTT)
Catalog # MOLE89914250-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:192.24 g/mol
CAS Number:21871-47-6
Formula:C₈H₈N₄S
MDL Number:MFCD00068731