Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Amino-4-bromo-5-methylisoxazole
3-Amino-4-bromo-5-methylisoxazole
3-Amino-4-bromo-5-methylisoxazole
3-Amino-4-bromo-5-methylisoxazole
Supplier:
Thermo Scientific
Certificates
Expand image
3-Amino-4-bromo-5-methylisoxazole
Supplier:
Thermo Scientific
About this item
CAS Number:
5819-40-9
:
Formula:
C₄H₅BrN₂O
MDL Number:
MFCD00052553
:
177.02 g/mol