0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
3-Amino-4-bromo-5-methylisoxazole
3-Amino-4-bromo-5-methylisoxazole
3-Amino-4-bromo-5-methylisoxazole
3-Amino-4-bromo-5-methylisoxazole
Supplier:
FLUOROCHEM
Certificates
Expand image
3-Amino-4-bromo-5-methylisoxazole
Supplier:
FLUOROCHEM
About this item
CAS Number:
5819-40-9
:
Formula:
C₄H₅BrN₂O
MDL Number:
MFCD00052553
:
177.02 g/mol