Lead(II) tetrafluoroborate 50% in aqueous solution
Catalog # HONE01502.9045
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pk:45 kg
- Seq:4388
Specifications
About this item
:
:380.81 g/mol
EINECS:237-486-0
CAS Number:13814-96-5
Formula:B₂F₈Pb
MDL Number:MFCD00016270