(±)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid hydrochloride, Sigma-Aldrich®
Catalog # SIAL214930-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:213.6608 g/mol
:M-Clarity™ quality level = MQ100
EINECS:255-610-1
CAS Number:41994-51-8
Formula:C₁₀H₁₂ClNO₂
MDL Number:MFCD00012743