Isoquinolin-3-ylmethanol
Catalog # COBBQA-2946-1G
Supplier: COMBI-BLOCKS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:159.1846 g/mol
CAS Number:76884-34-9
Formula:C₁₀H₉NO
MDL Number:MFCD01859798