Order Entry
Germany
ContactUsLinkComponent
1H-Tetrazole 0.40 M in acetonitrile
1H-Tetrazole 0.40 M in acetonitrile
Catalog # BI01551005-EA
CAS Number:  
1H-Tetrazole 0.40 M in acetonitrile
Catalog # BI01551005-EA
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :70.05 g/mol
EINECS:206-023-4
CAS Number:288-94-8
Formula:CH₂N₄
MDL Number:MFCD00005247