0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
1H-Tetrazole
1H-Tetrazole 0.45 mol/l in acetonitrile
1H-Tetrazole 0.45 mol/l in acetonitrile
:
Thermo Fisher Scientific
1H-Tetrazole 0.45 mol/l in acetonitrile
:
Thermo Fisher Scientific
:
288-94-8
:
:
CH₂N₄
:
MFCD00005247
:
70.05 g/mol
:
14,2786
:
0.798
:
Ambient
:
206-023-4
:
1648
:
3,II
:
GHS02
:
GHS07
msds_ALFAJ61034
Thermo Scientific Acroseal 25 Years