Order Entry
Germany
ContactUsLinkComponent
 
Buffers, potassium phosphate, 0,5 M solutions
 
Buffers, potassium phosphate, 0,5 M solutions
 

 

  • CAS No.:
    7778-77-0
  • MDL No.:
    MFCD00011401
  • Physical form:
    Liquid