Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer, EDTA buffer solution pH7,2 (0,5 mol/L)

Buffer, EDTA buffer solution pH7,2 (0,5 mol/L)

EDTA buffer solution pH7,2 (0,5 mol/l), Glass bottle, 2 mL
Catalog # UPST20-307
Marketsource
Supplier:   UPSTATE TECHNOLOGY
Buffer, EDTA buffer solution pH7,2 (0,5 mol/L)
Catalog # UPST20-307
Supplier:  UPSTATE TECHNOLOGY
Marketsource
CAS Number:  
Buffer, EDTA buffer solution pH7,2 (0,5 mol/L)

EDTA buffer solution pH7,2 (0,5 mol/l), Glass bottle, 2 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 2 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2 mL
  • Description:
    EDTA buffer solution pH7,2 (0,5 mol/l)

Specifications