ContactUsLinkComponent
Industrial gloves, GemoMech® 665
undefined
Industrial gloves, GemoMech® 665
  • Catalog No:
  • KCHL665.6
  • KCHL665.7
  • KCHL665.10
  • KCHL665.11
  • KCHL665.9
  • KCHL665.8
  • Model:
  • 665
  • 665
  • 665
  • 665
  • 665
  • 665
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Polyamide
  • Polyamide
  • Polyamide
  • Polyamide
  • Polyamide
  • Polyamide
  • Finish:
  • Dot coating on palm and fingertips
  • Dot coating on palm and fingertips
  • Dot coating on palm and fingertips
  • Dot coating on palm and fingertips
  • Dot coating on palm and fingertips
  • Dot coating on palm and fingertips
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388 (3121)
  • EN 388 (3121)
  • EN 388 (3121)
  • EN 388 (3121)
  • EN 388 (3121)
  • EN 388 (3121)