PEEKsil™ tubing, Upchurch Scientific®
PEEKsil capillary.
PEEKsil™ tubing is mechanically strong and has ideal characteristics for sealing with polymer fittings. It is comprised of high quality fused silica sheathed by PEEK tubing giving it excellent chemical compatibility. We hold it to very tight manufacturing tolerances delivering reliability. It can be an excellent replacement for stainless steel, PEEK, or standard fused silica.