Cleanroom overalls, 8R011
Supplier: FRISTADS KANSAS
Certificates
About this item
These overalls are made of XA32 fabric consisting of 99% polyester and 1% conductive fibres. The fibres are continuous filaament fibres, which means that the garments do not emit particles or fibres.
- Concealed zip and snap fasteners
- Crew neck with concealed snap fastener
- Elasticated cuffs, Raglan sleeves
- Open side legs
- Elasticated waistband with button holes
The carbon filaments produce good static discharge, thus preventing static charging and minimising triboelectric attraction of particles and fibres. This makes the garments very suitable for activities such as vehicle spraying.
Suitable for use in ISO class 7 cleanrooms.
Weight: 125 g/m²
Specifications
- Catalog No:
- KWIN100636540XL
- KWIN100636540S
- KWIN100636540XS
- KWIN100636540M
- KWIN100636540L
- KWIN1006365403XL
- KWIN1006365402XL
- Model:
- 8R011, XA32
- 8R011, XA32
- 8R011, XA32
- 8R011, XA32
- 8R011, XA32
- 8R011, XA32
- 8R011, XA32
- Garment application:
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Risk area:
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Weight:
- 125 g/m²
- 125 g/m²
- 125 g/m²
- 125 g/m²
- 125 g/m²
- 125 g/m²
- 125 g/m²