Test kit for cleaning control, GlowCheck
Product Details & Documents
GlowCheck offers users a method of visually checking cleaning and disinfection measures applied to surfaces by means of UV light.
- Very easy to use: The system can be used any time and anywhere
- Provides a quick overview of the effectiveness of the cleaning and disinfecting measures
- The marker can be used to add an additional note (date, time, etc.) below the stamp
- The special UV ink has been tested on a wide variety of surfaces and can usually be removed without leaving residues
- The saturated ink pad can be used for approx. 1 week before it must be re-saturated
Using an automatic stamp, the special UV-colour 'HYGIENE CHECK!' stamp is applied to the desired surface. The marks are not visible in daylight, but they shine brightly under the high-output 12-LED black-light torch. After cleaning, the black light torch helps determine how thoroughly the surface was cleaned or disinfected. If the mark is completely gone, the cleaning procedure was thorough. If all or part of the mark remains, then moist cleaning did not occur or was inadequate.
Delivery information: The system contains the following components:
GlowCheck special UV ink (30 ml) in a pump spray bottle
GlowCheck special UV stamp ink (30 ml)
GlowCheck marker
GlowCheck "HYGIENE CHECK!" automatic stamp
High-output 12-LED UV black-light torch
Training CD-ROM
Caution: As a precaution, a test should be performed on an inconspicuous area prior to use on critical materials (e.g. bare wood, old and porous plastic surfaces).
Under no circumstances must GlowCheck be used on medical devices such as ultrasonic transducers, etc.
Shake bottles well before use.
*The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- Description:Refill set for GlowCheck test kit