Pipette tips, HLT, HydroLogix™
Supplier: Thermo Fisher Scientific
Certificates
About this item
These tips feature a unique bi-fabricated wall that has been designed to produce a secure seal at a fraction of the force required to load standard pipette tips onto the pipettor. Accordingly, the forces required to eject the tips from your pipettor are significantly reduced. Molded from virgin polypropylene, they don't use additives or coatings that may affect sample integrity.
Limited Time Sale
Select items below are currently on sale
Specifications
- Catalog No:
- MLBP3800-11
- MLBP3895
- 613-7427
- 613-7433
- 613-7436
- 613-7431
- 613-7435
- 613-7426
- 613-7428
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)