Sealing film, self-adhesive for ELISA, PCR
211-0001
: Brand
- Optical property:Transparent
- Description:Sealing film, self-adhesive, PP, for PCR and ELISA, single sheets
- Seq:01
- Pk:100
Transparent PP film allows the visual inspection of the samples. Temperature range: −40 to +125 °C.