All chairs and stools are available with a star base which is made from high strength sectional steel tubing, which is particularly robust and durable. The flat design prevents trip hazards.
- Complete hygienic design, easy to clean and disinfect
- Seat height adjustment by gas-lift
- Seat tilt adjustment
- Backrest adjustable in height and inclination
Glides: The glides are wear-proof plastic and have a large surface area offering a high level of stability.
Castors: Dual-wheel castors with load-sensitive brakes have to be right for the floor. All chairs are supplied with soft castors for hard floors as standard. Hard castors for soft floors are also available as an option.
Stop&Go technology: For safety reasons chairs whose seat is more than 650 mm high must not be movable whilst in use. With innovative Stop&Go castors these chairs are only mobile when no one is sitting on them.
SoftTouch PU foam, resistant against aggressive liquids. The permanent contact backrest follows all movements automatically and supports dynamic sitting, ergonomical shaped backrest for ideal fitting to the back.
Conform to security level S2, GMP
- Catalog No:
- 132-0407
- 132-0406
- 132-0405
- 132-0498
- Environmentally Preferable:
- Sustainable Materials, Low Manufacturing Impact, Safer Human and Environmental Health
- Sustainable Materials, Low Manufacturing Impact, Safer Human and Environmental Health
- Sustainable Materials, Low Manufacturing Impact, Safer Human and Environmental Health
- Sustainable Materials, Low Manufacturing Impact, Safer Human and Environmental Health
- Environmental certification:
- ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
- ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
- ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
- ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)