ContactUsLinkComponent
Micro-splitter valves, Upchurch Scientific®
Micro-splitter valves, Upchurch Scientific®
Catalog # UPCHP-450
undefined
Micro-splitter valves, Upchurch Scientific®
Catalog # UPCHP-450
Specifications
  • Description:
    P-450 micro-splitter valve, medium pressure, 1/4-28 threads, biocompatible, 2,1 / 4,1 µl (closed / fully open) internal volume
  • Pk:
    1
Specifications
About this item
Designed to accurately split and control the low flow stream of a single incoming supply.
  • For interfacing LC-MS systems
  • Pressure rated to 800 psi (55 bar)
  • Adjustable split stream flow rates