ContactUsLinkComponent
Foam Pistol Bottle, Nito Clean
Foam Pistol Bottle, Nito Clean
Catalog # 141-0285
Supplier:  WERNER MERTZ
undefined
Foam Pistol Bottle, Nito Clean
Catalog # 141-0285
Supplier:  WERNER MERTZ
Specifications
  • Capacity:
    2,8 L
  • Seq:
    22
  • Pk:
    1
Specifications
About this item
Foam gun for applying a stable foam carpet on horizontal and vertical surfaces. The dosage can be adjusted.
For use with cleaning agents. Can be connected to a standard water tap.