Liquid soaps, Baktolin® sensitive
Supplier: BODE CHEMIE
Certificates
About this item
Baktolin® sensitive is a high quality liquid soap for the hands and skin. It is ideally suited for hand washing before surgical and after hygienic hand disinfectant. The nurturing components make Baktolin® sensitive particularly recommended for irritated and sensitive skin (for example, for patients and staff in clinics and doctors' surgeries, in care facilities or in laboratories and industrial kitchens).
- Gentle cleaning with moisturising and nurturing properties
- Very good compatibility for simultaneous use with alcohol-based hand disinfectants
- Bisabolol soothes skin irritation and inflammation
- Urea noticeably nourishes the skin and keeps it supple
- Neutral pH value of 5,5
- Free of alkali and soap, without pigments and colourants, discreetly scented
Caution: * These articles are only available in Germany.
*The product(s) on this page are outside the scope of the EU Directive 2019/904
*The product(s) on this page are outside the scope of the EU Directive 2019/904
Frequently Bought Together