ContactUsLinkComponent
Laboratory blenders, Seward Stomacher® 3500
Laboratory blenders, Seward Stomacher® 3500
Supplier:  Seward (FermionX)
Certificates
undefined
Laboratory blenders, Seward Stomacher® 3500
Supplier:  Seward (FermionX)
Specifications
  • Catalog No:
  • 432-0058
  • 432-0059
  • 432-3130
  • Model:
  • Stomacher® 3500
  • Stomacher® 3500
  • Stomacher® 3500
  • Paddle speed:
  • 75 - 180 min⁻¹ in steps of 5
  • 75 - 180 min⁻¹ in steps of 5
  • 75 - 180 min⁻¹ in steps of 5
  • Timer:
  • 0 – 99 min 59 s
  • 0 – 99 min 59 s
  • 0 – 99 min 59 s
  • W×D×H:
  • 470×450×470 mm
  • 470×450×470 mm
  • 470×450×470 mm
  • Weight:
  • 42 kg
  • 42 kg
  • 42 kg
Specifications