Safety shoes, slip-on, HACCP, X-Light, 711028/711029
Supplier: Abeba
Certificates
About this item
Anti-static shoes made from microfibre upper and lightweight, fuel resistant PU sole.
- Perspiration-absorbent lining with Silver Point
- Replaceable active comfort insole provides optimised comfort, orthopaedic insoles possible
- Sanitized® treatment reduces odour and bacterial growth
- Lightweight PU sole with high slip resistance and shock-absorbing system
- Protective steel toecap (200 J)
- Instep area with elastic band
- Washable 30 °C
Suitable for use in kitchens and in food-processing industries.
Colour: White/black
Certifications: DGUV 112-191; EN ISO 20345:2011; HACCP; S2, SRC