Occupational shoes, clogs, rubber, 9200 to 9310
: Abeba
Anti-static clogs made from perforated leather upper, a foldable, adjustable heel strap and a rubber sole with cross/circle profile, resistant to acids, bases (cleaning agents) and fuels.
- Leather lining
- Sanitised® treatment reduces odour and bacterial growth
- PU midsole
- Slip resistant special rubber sole
Colour: White, black or blue.
: EN ISO 20347:2012; OB, FO, SRC