About this item
Standard tips for general pipetting applications. Designed to fit most research-grade pipettors.
- Available with low binding surface technology for reduced sample retention
- Certified free from RNases, DNases, human DNA, PCR inhibitors and pyrogens
- XT tips (extended length): Ideal for pipetting from deep vessels
- LongReach, NX and type E tips: Ideal for molecular biology applications like PCR
- OneTouch tips create a leak proof seal: Easy to seal, easy to eject
Packaging options:
Racked
Sturdy racks that withstand several autoclave cycles
Cell Racks hold tips steadily in fully enclosed cells, minimise static and are excellent platforms for single and multichannel pipetting
Hinged racks hold the extremely popular 1000XT (50 to 1250 µl) tips, and traditional 1000 µl standard tips are placed in 100-hole standard racks
e•dek – Paperboard Rack System
Easy to recycle – compatible with most paperboard recycling programs
Used racks require minimal storage space – just flatten and recycle
Manufactured from paperboard sourced from sustainable forests
Printed with soy and vegetable based inks
Less weight equals reduced energy consumption and cost for shipping