Cut resistant gloves, HyFlex® 70-750
Supplier: Ansell
Certificates
About this item
These premium gloves use innovative technology for improved cut resistance, excellent flexibility and a higher level of comfort. Made from a combination of Kevlar®, Lycra® and stainless steel.
- Provide protection from heat and light molten splashes
- Intercept® cut resistance technology: Gloves are made from revolutionary engineered yarns with the highest cut-protection-to-weight ratio available
- Excellent flexibility and a high level of comfort
Reserved exclusively for the premium gloves, Ansell’s cut resistance technology uses revolutionary engineered yarns with the highest cut-protection-to-weight ratio available. This makes the HyFlex® gloves ideal for handling sharp tools and materials.
Ideal for assembling metal parts and components, handling sharp or irregular-edged metal parts, protection from heat & light molten splashes, welding and metal finish inspection.
Certifications: EN 388:2016 (2X4XD), EN 407 (X1XXXX); Cat. III
Specifications
- Catalog No:
- 112-3723
- 112-3726
- 112-3724
- 112-3725
- Model:
- 70-750
- 70-750
- 70-750
- 70-750
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Stainless steel/Lycra®/Kevlar®
- Stainless steel/Lycra®/Kevlar®
- Stainless steel/Lycra®/Kevlar®
- Stainless steel/Lycra®/Kevlar®
- Gauge:
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (2X4XD), EN 407 (X1XXXX)
- EN 388:2016 (2X4XD), EN 407 (X1XXXX)
- EN 388:2016 (2X4XD), EN 407 (X1XXXX)
- EN 388:2016 (2X4XD), EN 407 (X1XXXX)