ContactUsLinkComponent
General purpose gloves, Showa 7199NC
General purpose gloves, Showa 7199NC
  112-5893
 :  Showa
undefined
General purpose gloves, Showa 7199NC
  112-5893
 :  Showa
  • Glove size:
    10
  • Packed:
    12 pairs/pack
  • Ambidextrous / hand-specific:
    Hand-specific
  • Disposable / reusable:
    Reusable
  • Colour:
    Dark blue
  • Length:
    305 mm
  • Model:
    7199NC
  • EN norm:
    EN 388:2016 (4111X)
  • Liner material:
    Jersey
  • Finish:
    Smooth
  • Pk:
    72 PAIR
This flexible cotton jersey liner with full nitrile coating provides good mechanical resistance. Recommended for heavy-duty risks.
  • Absorbs perspiration, for comfortable extended wear
  • Protects the hand from oils and abrasion while retaining good touch
  • Impermeable for working in damp or greasy environments
  • Antibacterial and anti-odour treatment
  • Topstitched edge
Ideal for agriculture, automotive industry, food applications, glass and metallurgy, petrochemical industry and the public sector.
 : EN 388:2016 (4111X); Cat. II.