Chemical resistant gloves, Butoject® 898
Supplier: KCL by Honeywell
Certificates
About this item
These highly flexible, semi-disposable chemical protective gloves made of Butyl rubber are suitable for extreme demands when working with esters and ketones. Manufactured using the modern Vulcoject-process technology (injection moulding), this glove is particularly gas tight.
- Flexible at low temperatures, good ozone resistance: Can be used under extreme climatic conditions
- Very good protection against extremely aggressive chemicals
- High tightness against gas (meets NATO requirements)
- Powdered, smooth finish, rolled edge
- Wide shape makes it possible to wear gloves underneath
Certifications: EN 388 (2111X), EN ISO 374-1 Type A (BCIKLM), EN 421; Cat. III; AQL < 0,65
Specifications
- Catalog No:
- 112-4368
- 112-4367
- 112-1367
- 112-4369
- Model:
- 898
- 898
- 898
- 898
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Powdered / powder-free:
- Powdered
- Powdered
- Powdered
- Powdered
- Finish:
- Smooth
- Smooth
- Smooth
- Smooth
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- EN norm:
- EN 388 (2111X), EN ISO 374-1 Type A (BCIKLM), EN 421
- EN 388 (2111X), EN ISO 374-1 Type A (BCIKLM), EN 421
- EN 388 (2111X), EN ISO 374-1 Type A (BCIKLM), EN 421
- EN 388 (2111X), EN ISO 374-1 Type A (BCIKLM), EN 421