HPLC tubing, stainless steel, Upchurch Scientific®
Stainless steel.
- Seamless and pre-cut to ensure burr-free, flat connections
- Rockwell hardness (B) is maximum of 95
- Many sizes feature a colour-coded band for easy ID identification
- These products are RoHS compliant and meet ASTM A-269-10 and A-213
Each machine-cut end is polished, deburred on the inside and outside edges, and then the entire tubing is passivated. Finally, reagent-grade isopropanol is flushed through each piece. The thorough preparation and cleaning procedure guarantees tubing that is truly ready-to-use, with flat, burr-free ends and a clean finish. This care is important in achieving zero-dead-volume connections and good chromatographic results. Additionally, this tubing can be sterilised by numerous methods; however, if the tubing needs to be autoclaved, the colour-coded band (if present) should be removed beforehand.
: Stainless tubing are available in ¹/₁₆" and ¹/₈" outside diameter (OD) sizes, as well as 0,020" and ¹/₃₂" OD capillary sizes.