Disinfectant wipes, Bacillol® Tissues
Catalog # BODE980503
Supplier: BODE CHEMIE
New Product
Specifications
- Packed:100 wipes/dispenser, 1 dispenser/case
- Description:Bacillol® Tissues in dispenser
- Seq:02
- Pk:12
Specifications
About this item
Bacillol® Tissues are fragrance and aldehyde-free rapid disinfectant wipes for the convenient disinfection of alcohol resistant surfaces, such as work surfaces in medical practices, labs and clinics, and also medical equipment. The treated surfaces can be used after a period of just 30 seconds following drying.
- Can also be used without gloves thanks to the skin friendly formulation
- Easy dispensing from a practical, refillable dispenser
- Broad range of applications and material compatibility
- Bacillol® AF saturation fluid guarantees effective moistening
Microbiological effectiveness: bactericidal, yeasticidal, fungicidal, tuberculocidal, mycobactericidal, limited virucidal action (incl. HBV, HIV, HCV), MNV, adeno-, polyoma- and rotaviruses.
VAH, Robert Koch Institute (RKI) list of tested and recognised disinfectants and methods.
Packaging: 100 wipes per resealable, refillable dispenser. Wall mountings for the dispenser are also available.