Blender bags, BagLight®
Supplier: INTERSCIENCE
Certificates
About this item
These non-filter sample preparation bags are ideal for homogenising samples.
- Robust/ultra resistant
- Resistant to freezing
- Compatible with any blender
- Sterile and approved for food contact
BagLight® PolySilk®: Made of PolySilk® polyolefin complex, flexible and transparent. Available for different volume.
BagLight® Multilayer®: Made of Multilayer® reinforced complex, reinforced weldings.
BagLight® PolySilk® HD: Made of high density PolySilk®. They are rigid, matte and slightly opaque.