HPLC columns, BioBasic™
Supplier: Thermo Fisher Scientific
Certificates
About this item
The BioBasic™ HPLC columns are ideal for analysis of proteins, peptides and other biomolecules. The BioBasic reversed phase packings are available in C18, C8, C4, phenyl and cyano chemistries.