Sieve inserts for PULVERISETTE 15
Supplier: FRITSCH
Certificates
About this item
Sieve inserts determine the final fineness of a sample. These sieve inserts are available with trapezoidal or square perforations.
Sieve inserts with trapezoidal perforations improve size reduction through extra shearing action.
Sieve inserts with square perforations are used for the grinding of brittle materials and materials of medium fineness with a narrow grain band.