Order Entry
Germany
ContactUsLinkComponent
Magnesium nitrate solution Reag. Ph. Eur. 1049801
Flammable
Irritant
Magnesium nitrate solution Reag. Ph. Eur. 1049801
Catalog # 87862.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Irritant
Magnesium nitrate solution Reag. Ph. Eur. 1049801
Catalog # 87862.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
UN:1170
ADR:3,II
 :GHS02
 :GHS07
CAS Number:10377-60-3
 :13446-18-9
Formula:Mg(NO₃)₂