Order Entry
Germany
ContactUsLinkComponent
L(+)-Ascorbic acid Reag. Ph. Eur. 1008401
L(+)-Ascorbic acid Reag. Ph. Eur. 1008401
Catalog # 87782.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
L(+)-Ascorbic acid Reag. Ph. Eur. 1008401
Catalog # 87782.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :176.13 g/mol
UN:2265
ADR:3,III
 :GHS02
 :GHS08
 :GHS07
CAS Number:50-81-7
Formula:C₆H₈O₆
MDL Number:MFCD00064328