Methyl-2-methyl-5-nitrobenzoate
Catalog # COBBOS-1089-25G
Supplier: COMBI-BLOCKS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:195.18 g/mol
CAS Number:77324-87-9
Formula:C₉H₉NO₄
MDL Number:MFCD00230865