Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N,N'-Diethylhydrazine dihydrochloride
N,N'-Diethylhydrazine dihydrochloride ≥90%, tech.
N,N'-Diethylhydrazine dihydrochloride ≥90%, tech.
:
Thermo Fisher Scientific
N,N'-Diethylhydrazine dihydrochloride ≥90%, tech.
:
Thermo Fisher Scientific
:
7699-31-2
:
:
C₄H₁₄Cl₂N₂
:
MFCD00012523
:
159.06 g/mol
:
∼ 169 °C (dec.)
:
206-183-5
:
GHS06
:
GHS07
:
Ambient
msds_ALFAB22317
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years