(4-Fluorophenyl)methanesulphonyl chloride
Catalog # FLUO020883-1G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:208.64 g/mol
CAS Number:103360-04-9
Formula:C₇H₆ClFO₂S
MDL Number:MFCD01631930