Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Fluoro-o-anisidine hydrochloride
4-Fluoro-o-anisidine hydrochloride
4-Fluoro-o-anisidine hydrochloride
4-Fluoro-o-anisidine hydrochloride
Supplier:
COMBI-BLOCKS
Certificates
Expand image
4-Fluoro-o-anisidine hydrochloride
Supplier:
COMBI-BLOCKS
About this item
CAS Number:
178671-97-1
:
Formula:
C₇H₉ClFNO
MDL Number:
MFCD09800850
:
177.6000 g/mol