Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(4-tert-Butylphenyl)methanesulphonyl chloride
(4-tert-Butylphenyl)methanesulphonyl chloride
(4-tert-Butylphenyl)methanesulphonyl chloride
(4-tert-Butylphenyl)methanesulphonyl chloride
Supplier:
Thermo Scientific
Certificates
Expand image
(4-tert-Butylphenyl)methanesulphonyl chloride
Supplier:
Thermo Scientific
About this item
CAS Number:
519056-61-2
:
Formula:
C₁₁H₁₅ClO₂S
MDL Number:
MFCD04115413