Order Entry
Germany
ContactUsLinkComponent
4-(3-Chloro-5-fluorophenyl)-1-buten-4-ol
4-(3-Chloro-5-fluorophenyl)-1-buten-4-ol
Catalog # FLUO022226-1G
Supplier:  FLUOROCHEM
CAS Number:  
4-(3-Chloro-5-fluorophenyl)-1-buten-4-ol
Catalog # FLUO022226-1G
Supplier:  FLUOROCHEM
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
CAS Number:842123-72-2
Formula:C₁₀H₁₀ClFO
MDL Number:MFCD06201122