4-(4-Bromophenyl)-1-buten-4-ol
Catalog # FLUO022242-1G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
CAS Number:113842-31-2
Formula:C₁₀H₁₁BrO
MDL Number:MFCD06201137