Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromophenacyl trifluoromethanesulphonate
4-Bromophenacyl trifluoromethanesulphonate
4-Bromophenacyl trifluoromethanesulphonate
4-Bromophenacyl trifluoromethanesulphonate
Supplier:
BACHEM BIOCHEMICA
Certificates
Expand image
4-Bromophenacyl trifluoromethanesulphonate
Supplier:
BACHEM BIOCHEMICA
About this item
CAS Number:
93128-04-2
:
Formula:
C₉H₆BrF₃O₄S
MDL Number:
MFCD00467440