4-(4-Ethylphenyl)-1-buten-4-ol
Catalog # FLUO022235-1G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
CAS Number:221910-67-4
Formula:C₁₂H₁₆O
MDL Number:MFCD06201130