ContactUsLinkComponent
(Chloromethylene)dimethylammonium chloride 96%
Corrosive
Irritant
Health
(Chloromethylene)dimethylammonium chloride 96%
Certificates
undefined
Corrosive
Irritant
Health
(Chloromethylene)dimethylammonium chloride 96%
Specification Test Results
  • Appearance:
    Light yellow to light brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    95 to 105 %