ContactUsLinkComponent
Overpack protection cover, PIG®
Overpack protection cover, PIG®
Catalog # 121-0157
Supplier:  NEW PIG
undefined
Overpack protection cover, PIG®
Catalog # 121-0157
Supplier:  NEW PIG
Specifications
  • Ø×L:
    830×580 mm
  • Description:
    Overpack protective cover, UV resistant, for 360- and 246-litre overpack spill kits
  • Pk:
    1
Specifications
About this item
Durable cover adds UV protection and visibility to PIG overpack spill kit.
  • Durable PVC-laminated polyester cover
  • Elasticised bottom to fit PIG® spill kits in 246- and 360-litre overpacks
  • Keeps water, snow, dirt and debris off overpack lid and resists weathering and UV
  • Features 230×280 mm see-through pocket for identification label
  • High visibility colour to make kit easy to locate