Order Entry
Germany
ContactUsLinkComponent
Absorbent sock, PIG® oil-only
Absorbent sock, PIG® oil-only
Catalog # 121-0129
Supplier:  NEW PIG
undefined
Absorbent sock, PIG® oil-only
Catalog # 121-0129
Supplier:  NEW PIG

Specifications

  • Description:
    Oil-only socks (for indoor use)
  • Seq:
    01
  • Absorbency:
    113,7 l/box
  • Packed:
    30/box
  • Pk:
    30

Specifications

About this item

Cellulose filler and light brown coloured PP skin.

  • Mouldable socks hug corners and walls, surround machine bases and contain spills
  • Tan colour hides grime to stay on the job longer; discourages replacement before the sock is saturated
  • 95 % recycled content: Cellulose filler is made from newsprint

Hydrophobic cellusose filler for cleaning up spills of oils and oil based liquids.