Disinfectant, non sterile chlorine disinfectant, Zircon
A non foaming, sodium hypochlorite based, disinfectant.
- Ideal for use on walls, floors and equipment
- Suitable for use in CIP and spraywash enclosed systems
- Dilution for use: 0,5 to 2% v/v
Areas of use include walls, floors and equipment such as CIP and spraywash enclosed systems. Zircon is not soft metal safe, but is suitable for use on stainless steel and is also ideal for drain and mop disinfection.
All disinfectants are supported by comprehensive technical files which provide all the documentation for validation. All disinfectants are supported by the BPD and have independent efficacy data to prove performance.
: Supplied with its own certificate of analysis. A detailed technical file (including: product specification, manufacturing process, shelf life, microbiological and analytical data) is available on request.
: Available in 5, 20 and 1000 litre pack sizes.